Luminol (3-Aminophthalhydrazid) ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: NSC 5064

11050
AATB11050EA 120 EUR
AATB11050
Luminol (3-Aminophthalhydrazid) ≥95%
Luminol (3-Aminophthalhydrazid)

Luminol [3-Aminophthalhydrazide] is a luminescent peroxidase substrate.

Formel: C₈H₇N₃O₂
Molecular Weight: 177.16 g/mol
MDL: MFCD00006890
CAS-Nummer: 521-31-3
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR