Fluorescein tyramide

Lieferant: AAT BIOQUEST

11062
AATB11062EA 283 EUR
AATB11062
Fluorescein tyramide
Fluorescein tyramide

Fluorescein Tyramide is a green fluorescent reagent widely used for tyramide signal amplification (TSA) in IHC, ICC, FISH and multicolor FISH.


  • Stable for 12 months


Quality tested in-house


Bitte beachten For Research Use Only

Molecular Weight: 495,49 g/mol
CAS-Nummer: 210236-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR