Cy7 tyramide
Lieferant: AAT BIOQUEST
11064
AATB11064EA
283
EUR
AATB11064
Cy7 tyramide
Cy7 tyramide
Tyramide signal amplification (TSA) has proven to be a particularly versatile and powerful enzyme amplification technique with improved assay sensitivity.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only