XFD350 tyramide
Lieferant: AAT BIOQUEST
Achtung
11067
AATB11067EA 292 EUR
AATB11067
XFD350 tyramide
XFD350 tyramide
XFD350 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 350 (Alexa Fluor® is the trademark of ThermoFisher).
  • Bright signal
  • Excellent signal amplification for low expression targets
ADR: 5,II