XFD488 tyramide
Lieferant: AAT BIOQUEST
|
Achtung
|
11070
AATB11070EA
307
EUR
AATB11070
XFD488 tyramide
XFD488 tyramide
XFD488 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher).
- Bright signal
- Excellent signal amplification for low expression targets
ADR:
5,II |