XFD514 tyramide
Lieferant: AAT BIOQUEST
11071
AATB11071EA
283
EUR
AATB11071
XFD514 tyramide
XFD514 tyramide
XFD 514 tyramide contains the Alexa Fluor® 514 fluorophore that can be readily detected with the standard rhodamine 6G filter set (Alexa Fluor® is the trademark of ThermoFisher).
- Stable for 12 months
Tyramide signal amplification (TSA) has proven to be a particularly versatile and powerful enzyme amplification technique with improved assay sensitivity.
Bitte beachten For Research Use Only