XFD532 tyramide
Lieferant: AAT BIOQUEST
11072
AATB11072EA
283
EUR
AATB11072
XFD532 tyramide
XFD532 tyramide
XFD 532 tyramide contains the Alexa Fluor® 532 fluorophore that can be readily detected with the Alexa Fluor® 532/ATTO 532 filter set (Alexa Fluor® is the trademark of ThermoFisher).
- Stable for 12 months
Tyramide signal amplification (TSA) has proven to be a particularly versatile and powerful enzyme amplification technique with improved assay sensitivity.
Bitte beachten For Research Use Only