XFD546 tyramide
Lieferant: AAT BIOQUEST
Achtung
11075
AATB11075EA 307 EUR
AATB11075
XFD546 tyramide
XFD546 tyramide
XFD546 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 546 (Alexa Fluor® is the trademark of ThermoFisher).
  • Bright signal
  • Excellent signal amplification for low expression targets
ADR: 5,II