XFD546 tyramide
Lieferant: AAT BIOQUEST
|
Achtung
|
11075
AATB11075EA
307
EUR
AATB11075
XFD546 tyramide
XFD546 tyramide
XFD546 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 546 (Alexa Fluor® is the trademark of ThermoFisher).
- Bright signal
- Excellent signal amplification for low expression targets
ADR:
5,II |