XFD568 tyramide
Lieferant: AAT BIOQUEST
Achtung
11078
AATB11078EA 292 EUR
AATB11078
XFD568 tyramide
XFD568 tyramide
XFD568 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 568 (Alexa Fluor® is the trademark of ThermoFisher).
  • Bright signal
  • Excellent signal amplification for low expression targets
ADR: 5,II