XFD594 tyramide
Lieferant: AAT BIOQUEST
Achtung
11082
AATB11082EA 307 EUR
AATB11082
XFD594 tyramide
XFD594 tyramide
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
  • Bright signal
  • Excellent signal amplification for low expression targets
ADR: 5,II