Amplite® Fluorimetric Fluorescamine Protein Quantitation Kit *Blue Fluorescence*
Lieferant: AAT BIOQUEST
11100
AATB11100EA 307 EUR
AATB11100
Amplite® Fluorimetric Fluorescamine Protein Quantitation Kit *Blue Fluorescence*
Tests Enzymtests
Fluorescamine is intrinsically non-fluorescent but reacts rapidly with primary aliphatic amines, including those in peptides and proteins, to yield a blue-green-fluorescent derivative.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Bitte beachten For research use only.