ReadiLink™ Rapid mFluor™ Violet 540 Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Lieferant: AAT BIOQUEST
1114
AATB1114EA 230 EUR
AATB1114
ReadiLink™ Rapid mFluor™ Violet 540 Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Proteinkonjugate und Markierungen
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
  • Violet-laser excitable
  • Robust and simple antibody labelling protocol
  • No column purification required
Bitte beachten For research use only.