ReadiLink™ Rapid mFluor™ Violet 610 Antibody Labelling Kit
Lieferant: AAT BIOQUEST
1116
AATB1116EA
232
EUR
AATB1116
ReadiLink™ Rapid mFluor™ Violet 610 Antibody Labelling Kit
Proteinkonjugate und Markierungen
Microscale optimised for labeling 50 µg antibody per reaction. ReadiLink™ Rapid mFluor™ Violet 610 antibody labelling kits provide a quick and convenient method to label antibodies, proteins (<10 kDa), or other amine-containing biomolecules.
- Robust protocol
- Consistent results
- 12-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry and microscopy.
Lieferumfang: Shipped at RT.
Bitte beachten Research use only.