ReadiLink™ Rapid mFluor™ Blue 570 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Lieferant: AAT BIOQUEST
1120
AATB1120EA 230 EUR
AATB1120
ReadiLink™ Rapid mFluor™ Blue 570 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Proteinkonjugate und Markierungen
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
  • Blue-laser excitable
  • Robust and simple antibody labelling protocol
  • No column purification required
Bitte beachten For research use only.