Amplite® Colorimetric Glucose Oxidase Assay Kit
Lieferant: AAT BIOQUEST
11299
AATB11299EA 460 EUR
AATB11299
Amplite® Colorimetric Glucose Oxidase Assay Kit
Tests Enzymtests
The glucose oxidase is a dimeric protein that catalyses the oxidation of beta-D-glucose into hydrogen peroxide and D-glucono-1,5-lactone, which is hydrolysed to gluconic acid.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Bitte beachten For research use only.