Amplite® Colorimetric Glucose Oxidase Assay Kit
Lieferant: AAT BIOQUEST
11299
AATB11299EA
460
EUR
AATB11299
Amplite® Colorimetric Glucose Oxidase Assay Kit
Tests
Enzymtests
The glucose oxidase is a dimeric protein that catalyses the oxidation of beta-D-glucose into hydrogen peroxide and D-glucono-1,5-lactone, which is hydrolysed to gluconic acid.
- In-house QC testing
- Sensitive
- Suggested protocol available
Bitte beachten For research use only.