Amplite® Fluorimetric Myeloperoxidase Assay Kit *Red Fluorescence*
Lieferant: AAT BIOQUEST
11301
AATB11301EA
307
EUR
AATB11301
Amplite® Fluorimetric Myeloperoxidase Assay Kit *Red Fluorescence*
Tests
Enzymtests
Myeloperoxidase (MPO), most abundantly present in neutrophils and monocytes, is a green hemoprotein having peroxidase activity.
- In-house QC testing
- Sensitive
- Suggested protocol available
Bitte beachten For research use only.