Amplite® Colorimetric Xanthine Oxidase Assay Kit
Lieferant: AAT BIOQUEST
11307
AATB11307EA 307 EUR
AATB11307
Amplite® Colorimetric Xanthine Oxidase Assay Kit
Tests Enzymtests
Xanthine oxidase (XO)is an enzyme that catalyzes the oxidation of hypoxanthine to xanthine and can further catalyse the oxidation of xanthine to uric acid.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Bitte beachten For research use only.