Amplite® Fluorimetric Neutrophil Elastase Activity Assay Kit
Lieferant: AAT BIOQUEST
11327
AATB11327EA 538 EUR
AATB11327
Amplite® Fluorimetric Neutrophil Elastase Activity Assay Kit
Tests Enzymtests
Designed for consistent and dependable results. The Amplite® fluorimetric neutrophil elastase activity assay kit provides a robust and accurate method for quantifying neutrophil elastase (NE) activity.
  • Ex/Em= 360/470 nm
  • Consistent results
  • Six-month shelf life
The kit’s straightforward protocol is optimised for the sensitive detection of NE in blood and plasma samples and is suitable for screening and characterising chemotactic agents triggering neutrophil stimulation.
Lieferumfang: Shipped with blue ice.
Bitte beachten Research use only.