Amplite® Fluorimetric Acetylcholinesterase Assay Kit *Red Fluorescence*
Lieferant: AAT BIOQUEST
11402
AATB11402EA 307 EUR
AATB11402
Amplite® Fluorimetric Acetylcholinesterase Assay Kit *Red Fluorescence*
Tests Enzymtests
Acetylcholinesterase, also known as AChE, is an enzyme that degrades (through its hydrolytic activity) the neurotransmitter acetylcholine, producing choline and an acetate group.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Bitte beachten For research use only.