Amplite® Fluorimetric Acetylcholine Assay Kit *Red Fluorescence*
Lieferant: AAT BIOQUEST
11403
AATB11403EA 307 EUR
AATB11403
Amplite® Fluorimetric Acetylcholine Assay Kit *Red Fluorescence*
Tests Enzymtests
Acetylcholine and its metabolites are needed for three main physiological purposes: structural integrity and signaling roles for cell membranes, cholinergic neurotransmission (acetylcholine synthesis), and as a major source for methyl groups via its metabolite, trimethylglycine (betaine) that participates in the S-adenosylmethionine synthesis pathways.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Bitte beachten For research use only.