Violet 540 acid, mFluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1142
AATB1142EA 460 EUR
AATB1142
Violet 540 acid, mFluor™ fluoreszierende Probe
Violet 540 acid

mFluor™ dyes are developed for multicolor flow cytometry-focused applications.


  • pH-insensitive
  • High photostability
  • Brighter signal

Molecular Weight: 1600 kDa
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa5d50b1707a4fff4d0ea7badeb6a65f25d6b4e0&vid=02b8daef8be6205beffc028288be7720cacbede2&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR