Blue 570 acid, mFluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
1143
AATB1143EA
460
EUR
AATB1143
Blue 570 acid, mFluor™ fluoreszierende Probe
Blue 570 acid
mFluor™ Blue 570 dyes are an excellent alternative to RPE since they have the spectral properties equivalent to those of RPE conjugates.
- pH-insensitive
- High photostability
- Brighter signal
Molecular Weight:
1433,37 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...