Green 620 acid, mFluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1144
AATB1144EA 460 EUR
AATB1144
Green 620 acid, mFluor™ fluoreszierende Probe
Green 620 acid

mFluor™ Green 620 dye is a unique dye that is well excited by the green laser at 532 nm to give red fluorescence.


  • pH-insensitive
  • High photostability
  • Brighter signal

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23edc15edeb4e4c2f11b02fcdf2a27125c3b5c2f&vid=d95dcc066736b86dfc39f72c4d9d1d6b35b288c7&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR