Red 780 acid, mFluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1147
AATB1147EA 460 EUR
AATB1147
Red 780 acid, mFluor™ fluoreszierende Probe
Red 780 acid

mFluor™ Red 780 dyes are an excellent alternative to APC-Alexa Fluor® 750 tandems since they have the spectral properties equivalent to those of APC-Alexa Fluor® 750 conjugates.


  • pH-insensitive
  • High photostability
  • Brighter signal

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=598e42d502174b0bf5f56f912ddce5d05a367762&vid=42be6bc17ee786e02569469f4935c53ffcfd9235&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR