Violet 450 succinimidyl ester, mFluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Violet 450 SE

1150
AATB1150EA 307 EUR
AATB1150
Violet 450 succinimidyl ester, mFluor™ fluoreszierende Probe
Violet 450 succinimidyl ester

AAT Bioquest's mFluor™ dyes are developed for multicolor flow cytometry-focused applications.


  • pH-insensitive
  • High photostability
  • Brighter signal

Molecular Weight: 841,96 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR