Violet 510 succinimidyl ester, mFluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Violet 510 SE
1151
AATB1151EA
307
EUR
AATB1151
Violet 510 succinimidyl ester, mFluor™ fluoreszierende Probe
Violet 510 succinimidyl ester
AAT Bioquest's mFluor™ dyes are developed for multicolor flow cytometry-focused applications.
- pH-insensitive
- High photostability
- Brighter signal
Molecular Weight:
770,83 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...