Violet 540 succinimidyl ester, mFluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Violet 540 SE
1152
AATB1152EA
307
EUR
AATB1152
Violet 540 succinimidyl ester, mFluor™ fluoreszierende Probe
Violet 540 succinimidyl ester
AAT Bioquest's mFluor™ dyes are developed for multicolor flow cytometry-focused applications.
- pH-insensitive
- High photostability
- Brighter signal
Molecular Weight:
1535,88 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...