mFluor™ Violet 550 SE
Lieferant: AAT BIOQUEST
1153
AATB1153EA 283 EUR
AATB1153
mFluor™ Violet 550 SE
mFluor™ Violet 550 SE
mFluor™ Violet 550 dyes have fluorescence excitation and emission maxima of ~405 nm and ~550 nm respectively.
  • Stable for 12 months
mFluor™ dyes are developed for multicolor flow cytometry-focused applications.
Bitte beachten For Research Use Only