mFluor™ Violet 590 SE
Lieferant: AAT BIOQUEST
1155
AATB1155EA
283
EUR
AATB1155
mFluor™ Violet 590 SE
mFluor™ Violet 590 SE
mFluor™ Violet 590 dyes have fluorescence excitation and emission maxima of ~405 nm and ~590 nm respectively
- Stable for 12 months
mFluor™ dyes are developed for multicolor flow cytometry-focused applications.
Bitte beachten For Research Use Only