Violet 610 succinimidyl ester, mFluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Violet 610 SE

1156
AATB1156EA 292 EUR
AATB1156
Violet 610 succinimidyl ester, mFluor™ fluoreszierende Probe
Violet 610 succinimidyl ester

Advances in spectral flow cytometers have expanded applications and capabilities beyond conventional flow cytometry.


  • pH-insensitive
  • High photostability
  • Brighter signal

Molecular Weight: 1293,46 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR