mFluor™ Violet 480 SE
Lieferant: AAT BIOQUEST
1158
AATB1158EA 283 EUR
AATB1158
mFluor™ Violet 480 SE
mFluor™ Violet 480 SE
mFluor™ Violet 480 SE is reasonably stable and shows good reactivity and selectivity with protein amino groups.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only