mFluor™ Blue 585 SE
Lieferant: AAT BIOQUEST
1159
AATB1159EA
464
EUR
AATB1159
mFluor™ Blue 585 SE
mFluor™ Blue 585 SE
mFluor™ Blue 585 dyes are an excellent alternative to RPE since they have the spectral properties equivalent to those of RPE conjugates.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only