Blue 590 succinimidyl ester, mFluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Blue 590 SE
1161
AATB1161EA
439
EUR
AATB1161
Blue 590 succinimidyl ester, mFluor™ fluoreszierende Probe
Blue 590 succinimidyl ester
mFluor™ Blue 590 dye can be well excited with blue laser at 488 nm.
- pH-insensitive
- High photostability
- Brighter signal
Molecular Weight:
1783,14 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...