4-Nitrophenyl phosphate disodium salt (pNNP) ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Phosphoric acid 4-nitrophenyl ester disodium salt, 4-Nitrophenyl phosphate disodium salt, PNPP disodium, pNNP
11619
AATB11619EA
120
EUR
AATB11619
4-Nitrophenyl phosphate disodium salt (pNNP) ≥95%
4-Nitrophenyl phosphate disodium salt (pNNP)
para-Nitrophenylphosphate (pNPP) is a chromogenic substrate for acid and alkaline phosphatase in ELISA assays.
Formel:
C₆H₄NNa₂O₆P Molecular Weight: 263,052 g/mol |
MDL:
MFCD00007319 CAS-Nummer: 4264-83-9 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...