mFluor™ Blue 615 SE
Lieferant: AAT BIOQUEST
1162
AATB1162EA 464 EUR
AATB1162
mFluor™ Blue 615 SE
mFluor™ Blue 615 SE
mFluor™ Blue 615 dye can be well excited with blue laser at 488 nm.
  • Stable for 24months
Quality tested in-house
Bitte beachten For Research Use Only