Blue 620 succinimidyl ester, mFluor™ fluoreszierende Probe
Lieferant: AAT Bioquest
|
Achtung
|
1163
AATB1163EA
439
EUR
AATB1163
Blue 620 succinimidyl ester, mFluor™ fluoreszierende Probe
Blue 620 succinimidyl ester
mFluor™ Blue 620 dye can be well excited with blue laser at 488 nm.
- pH-insensitive
- High photostability
- Brighter signal
ADR:
5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...