PhosLite™ Green
Lieferant: AAT BIOQUEST
Achtung
11630
AATB11630EA 307 EUR
AATB11630
PhosLite™ Green
PhosLite™ Green
This water-soluble PhosLite™ Green substrate generates a bright and photostable yellow-green fluorescent precipitate at the site of phosphtase activity.
  • Extremely large Stokes shift
  • Bright and stable signal
  • High photostability
ADR: 5,II