PhosLite™ Green
Lieferant: AAT BIOQUEST
|
Achtung
|
11630
AATB11630EA
307
EUR
AATB11630
PhosLite™ Green
PhosLite™ Green
This water-soluble PhosLite™ Green substrate generates a bright and photostable yellow-green fluorescent precipitate at the site of phosphtase activity.
- Extremely large Stokes shift
- Bright and stable signal
- High photostability
ADR:
5,II |