Blue 630 succinimidyl ester, mFluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Blue 630 SE
1164
AATB1164EA
439
EUR
AATB1164
Blue 630 succinimidyl ester, mFluor™ fluoreszierende Probe
Blue 630 succinimidyl ester
mFluor™ Blue 630 dye can be well excited with blue laser at 488 nm.
- pH-insensitive
- High photostability
- Brighter signal
Molecular Weight:
709,86 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...