Resorufin Phosphocholine
Lieferant: AAT BIOQUEST
Achtung
11640
AATB11640EA 460 EUR
AATB11640
Resorufin Phosphocholine
Resorufin Phosphocholine
Glycerophosphocholine phosphodiesterase release phosphocholine from resorufin phosphocholine to give the highly fluorescent resorufin that can be readily monitored by either fluorescence or absorption.
  • Bright signal
  • Adaptable to multiple applications
ADR: 5,II