Resorufin Phosphocholine
Lieferant: AAT BIOQUEST
|
Achtung
|
11640
AATB11640EA
460
EUR
AATB11640
Resorufin Phosphocholine
Resorufin Phosphocholine
Glycerophosphocholine phosphodiesterase release phosphocholine from resorufin phosphocholine to give the highly fluorescent resorufin that can be readily monitored by either fluorescence or absorption.
- Bright signal
- Adaptable to multiple applications
ADR:
5,II |