Green 620 succinimidyl ester, mFluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Green 620 SE

1165
AATB1165EA 460 EUR
AATB1165
Green 620 succinimidyl ester, mFluor™ fluoreszierende Probe
Green 620 succinimidyl ester

mFluor™ Green 620 dye is a unique dye that is well excited by the green laser at 532 nm to give red fluorescence.


  • pH-insensitive
  • High photostability
  • Brighter signal

Molecular Weight: 883,98 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR