mFluor™ Green 615 SE
Lieferant: AAT BIOQUEST
1166
AATB1166EA 442 EUR
AATB1166
mFluor™ Green 615 SE
mFluor™ Green 615 SE
mFluor™ Green 615 dye can be well excited with green laser at 532 nm.
  • Stable for 24months
Quality tested in-house
Bitte beachten For Research Use Only