Green 630 succinimidyl ester, mFluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
1168
AATB1168EA
439
EUR
AATB1168
Green 630 succinimidyl ester, mFluor™ fluoreszierende Probe
Green 630 succinimidyl ester
Advances in spectral flow cytometers have expanded applications and capabilities beyond conventional flow cytometry.
- pH-insensitive
- High photostability
- Brighter signal
ADR:
5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...