Blue 580 succinimidyl ester, mFluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Blue 580 SE

1178
AATB1178EA 439 EUR
AATB1178
Blue 580 succinimidyl ester, mFluor™ fluoreszierende Probe
Blue 580 succinimidyl ester

Advances in spectral flow cytometers have expanded applications and capabilities beyond conventional flow cytometry.


  • pH-insensitive
  • High photostability
  • Brighter signal

Molecular Weight: 832,98 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc52b98616cf2fd11cbaa8bce7cfff34e8cf7e13&vid=c78607011283d605ce019c9dd1b59d73fe689486&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR