mFluor™ Blue 583 SE
Lieferant: AAT BIOQUEST
1179
AATB1179EA
464
EUR
AATB1179
mFluor™ Blue 583 SE
mFluor™ Blue 583 SE
mFluor™ Blue 583 dyes are water-soluble, and the protein conjugates prepared with mFluor™ Blue 583 dyes are well excited at 488 nm to give bright red fluorescence.
- Stable for 12 months
AAT Bioquest's mFluor™ dyes are developed for multicolor flow cytometry-focused applications, in particular, for spectral fluorescence flow cytometry.
Bitte beachten For Research Use Only