Amplite® Fluorimetric Asparaginase Activity Assay Kit
Lieferant: AAT BIOQUEST
11799
AATB11799EA
673
EUR
AATB11799
Amplite® Fluorimetric Asparaginase Activity Assay Kit
Tests
Enzymtests
Designed for consistent and dependable results. The Amplite® Fluorimetric Asparaginase Activity Assay Kit provides a simple and straightforward procedure for measuring Asparaginase activity in a variety of biological samples.
- Ex/Em=540/590 nm
- Consistent results
- Six-month shelf life
One unit (U) is the amount of enzyme that catalyzes the reaction of 1 µmol of substrate per minute.
Lieferumfang: Shipped with blue ice.
Bitte beachten Research use only.