5-FAM-X, SE ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 6-(Fluorescein-5-carboxamido)hexanoic acid, succinimidyl ester, 5-FAM-X, NHS ester

119
AATB119EA 152 EUR
AATB119
5-FAM-X, SE ≥95%
5-FAM-X, SE

5-FAM-X, SE contains the aminohexanoyl spacer (known as 'X') between the FAM fluorophore and the succinimidyl ester.

Formel: C₃₁H₂₆N₂O₁₀
Molecular Weight: 586,55 g/mol
CAS-Nummer: 148356-00-7
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR