Red 780 amine, mFluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1210
AATB1210EA 460 EUR
AATB1210
Red 780 amine, mFluor™ fluoreszierende Probe
Red 780 amine

mFluor™ Red 780 dyes are an excellent alternative to APC-Alexa Fluor® 750 tandems since they have the spectral properties equivalent to those of APC-Alexa Fluor® 750 conjugates.


  • pH-insensitive
  • High photostability
  • Brighter signal

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cfea683f8cc6ac1b49f576325d365d1a6b095aa&vid=e70bda4739718bbea48c4c8647cb1c87cb2d10b2&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR