D(-)-Luciferin ≥95%

Lieferant: AAT Bioquest
Achtung

12501 12502 12503
AATB12501EA 120 EUR
AATB12501 AATB12502 AATB12503
D(-)-Luciferin ≥95%
D(-)-Luciferin

Luciferin is the most popular and versatile bioluminescent substrate.

Formel: C₁₁H₈N₂O₃S₂
MDL: MFCD00042929
CAS-Nummer: 2591-17-5
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5d04b52b6545c43aed5376841e481c2729965b3&vid=437b0f0187e0e5be2eb461c20b7cb8f28de3be0c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR