D(-)-Luciferin Kaliumsalz ≥95%
Lieferant: AAT Bioquest
|
Achtung
|
12505
12506
12507
AATB12505EA
120
EUR
AATB12505
AATB12506
AATB12507
D(-)-Luciferin Kaliumsalz ≥95%
D(-)-Luciferin Kaliumsalz
Luciferin is the most popular and versatile bioluminescent substrate.
Formel:
C₁₁H₇KN₂O₃S₂ |
MDL:
MFCD00044928 CAS-Nummer: 115144-35-9 ADR: 5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...