D-Luciferin 6′-O-phosphate trisodium salt ≥90%

Lieferant: AAT BIOQUEST
Achtung

12512
AATB12512EA 307 EUR
AATB12512
D-Luciferin 6′-O-phosphate trisodium salt ≥90%
D-Luciferin 6′-O-phosphate trisodium salt

D-Luciferin phosphate is a highly sensitive bioluminescent substrate for detecting alkaline phosphatases and protein phosphatases by luminometric detection.

Formel: C₁₁H₆N₂Na₃O₆PS₂
Molecular Weight: 426,25 g/mol
MDL: MFCD00214322
CAS-Nummer: 145613-12-3
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR